Adenosin-5'-triphosphat Dinatriumsalz (ATP Dinatriumsalz)
Lieferant: PanReac AppliChem
A1348.0100
A1348.0100EA
404
EUR
A1348.0100
Adenosin-5'-triphosphat Dinatriumsalz (ATP Dinatriumsalz)
Adenosin-5'-triphosphat Dinatriumsalz (ATP Dinatriumsalz)
Formel:
C₁₀H₁₄N₅Na₂O₁₃P₃ |
MDL:
MFCD00080339 CAS-Nummer: 987-65-5 EINECS: 213-579-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...