D(+)-Glucose Monohydrat

Lieferant: PanReac AppliChem

Synonyme: Dextrose Monohydrat, Traubenzucker Monohydrat

A1349.5000
A1349.5000EA 128 EUR
A1349.5000
D(+)-Glucose Monohydrat
D(+)-Glucose Monohydrat
Formel: C₆H₁₂O₆·H₂O
Molecular Weight: 198,17 g/mol
Schmelzpunkt: 83 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00149450
CAS-Nummer: 14431-43-7
EINECS: 200-075-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9ea2af2ca62ca94d86809070593a60aafd8cd1b&vid=e549b499dca7fb95811640efad1429268c8f6d8b&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR