2-Aminobenzothiazol 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Benzothiazol-2-ylamin, Cer(III)-Ionophor II

A13521.22 A13521.36
A13521.22EA 52.4 EUR
A13521.22 A13521.36
2-Aminobenzothiazol 98%
2-Aminobenzothiazole

Purity: 98% CAS No.: 136-95-8 Molecular Formula: C7H6N2S Formula Weight: 150.20 Melting Point: 126-129° Beilstein Reference: 116315 Merck Reference: 14,424 MDL No.: MFCD00005785

Formel: C₇H₆N₂S
Molecular Weight: 150.2 g/mol
Siedepunkt: 190…195 °C (0.05 mmHg)
Schmelzpunkt: 126…129 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005785
CAS-Nummer: 136-95-8
EINECS: 205-268-4
Merck Index: 13,00423

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR