3,5-Dichlorbenzylalkohol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A13527.06 A13527.14
A13527.06EA 43.8 EUR
A13527.06 A13527.14
3,5-Dichlorbenzylalkohol ≥98%
3,5-Dichlorbenzylalkohol

Purity: 98% CAS No.: 60211-57-6 Molecular Formula: C7H6Cl2O Formula Weight: 177.03 Melting Point: 76-80° Beilstein Reference: 1936389

Formel: C₇H₆Cl₂O
Siedepunkt: 264 °C (1013 hPa)
Schmelzpunkt: 76…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004634
CAS-Nummer: 60211-57-6
EINECS: 262-109-1
UN: 2250
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR