Benzylbromid ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99%
CAS No.: 100-39-0
Molecular Formula: C7H7Br
Formula Weight: 171.04
Flash Point: 87°(189°F)
Physical Form: Liquid
Density: 1.438
Refractive Index: 1.5750
Sensitivity: Moisture Sensitive
Odour: Pungent, makes eyes water
Melting Point: -4°
Boiling Point: 198-199°
Beilstein Reference: 385801
Merck Reference: 14,1128
UN No.: 1737
MDL No.: MFCD00000172
Formel:
(C₆H₅)CH₂Br Siedepunkt: 198…199 °C Schmelzpunkt: –4 °C Dichte: 1,438 Flammpunkt: 87 °C (189 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000172 CAS-Nummer: 100-39-0 EINECS: 202-847-3 UN: 1737 ADR: 6.1,II Merck Index: 14,1128 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...