trans-Zimtsäure ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 99+%
CAS No.: 140-10-3
Molecular Formula: C9H8O2
Formula Weight: 148.16
Flash Point: 166°(330°F)
Physical Form: Powder
Density: 1.250
Appearance: White to pale cream
Odour: Weak, characteristic
Melting Point: 133-136°
Boiling Point: 147°/4mm
Beilstein Reference: 1905952
Merck Reference: 14,2299
MDL No.: MFCD00004369
Formel:
C₉H₈O₂ Siedepunkt: 147 °C (4 mmHg) Schmelzpunkt: 133…136 °C Dichte: 1,250 Flammpunkt: 166 °C (330 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004369 CAS-Nummer: 140-10-3 EINECS: 205-398-1 Merck Index: 14,2299 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...