N,N-Dimethylformamid ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
DMF, Ameisensäuredimethylamid, N,N-Dimethylmethansäureamid
A13547.0D
A13547.0F
A13547.AK
A13547.0DEA
217
EUR
A13547.0D
A13547.0F
A13547.AK
N,N-Dimethylformamid ≥99%
N,N-Dimethylformamid
Purity: 99%
CAS No.: 68-12-2
Molecular Formula: HCON(CH3)2
Formula Weight: 73.09
Flash Point: 57°(135°F)
Refractive Index: 1.4310
Odour: Amine-like
pH: 7
Melting Point: -61°
Boiling Point: 153°
Beilstein Reference: 605365
Merck Reference: 14,3243
UN No.: 2265
MDL No.: MFCD00003284
Formel:
HCON(CH₃)₂ Molecular Weight: 73.09 g/mol Siedepunkt: 153 °C Schmelzpunkt: –61 °C Dichte: 0.944 Flammpunkt: 57 °C (135 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003284 CAS-Nummer: 68-12-2 EINECS: 200-679-5 UN: 2265 ADR: 3 III REACH: 01-2119475605-32 Merck Index: 13,03269 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...