4-(4-Nitrobenzyl)pyridin ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: 4-(p-Nitrobenzyl)Pyridine

A13564.06 A13564.14 A13564.22
A13564.06EA 38.5 EUR
A13564.06 A13564.14 A13564.22
4-(4-Nitrobenzyl)pyridin ≥98%
4-(4-Nitrobenzyl)pyridin

Purity: 98+% CAS No.: 1083-48-3 Molecular Formula: C12H10N2O2 Formula Weight: 214.22 Melting Point: 69-72° Beilstein Reference: 170877

Formel: C₁₂H₁₀N₂O₂
Molecular Weight: 214.22 g/mol
Schmelzpunkt: 69…72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006445
CAS-Nummer: 1083-48-3
EINECS: 214-108-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR