2,4-Dihydroxypyridin ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

A13566.03 A13566.06 A13566.14
A13566.03EA 48.7 EUR
A13566.03 A13566.06 A13566.14
2,4-Dihydroxypyridin ≥97%

Purity: 97% CAS No.: 626-03-9 Molecular Formula: C5H5NO2 Formula Weight: 111.10 Melting Point: ca 267° dec. Beilstein Reference: 108533

Formel: C₅H₅NO₂
Siedepunkt: 315 °C (1013 hPa)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006273
CAS-Nummer: 626-03-9
EINECS: 210-924-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR