9-(Chlormethyl)anthracen ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

A13579.03 A13579.06 A13579.14
A13579.03EA 35.8 EUR
A13579.03 A13579.06 A13579.14
9-(Chlormethyl)anthracen ≥98%
9-(Chloromethyl)anthracene

Purity: 98+% CAS No.: 24463-19-2 Molecular Formula: C15H11Cl Formula Weight: 226.71 Melting Point: 138-142° Beilstein Reference: 1873394 UN No.: 3261 MDL No.: MFCD00001263

Formel: C₁₅H₁₁Cl
Molecular Weight: 226.71 g/mol
Siedepunkt: 371
Schmelzpunkt: 138…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001263
CAS-Nummer: 24463-19-2
EINECS: 246-274-7
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR