o-Toluidin ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2-Methylanilin, 2-Aminotoluol, 2-Toluidin

A13602.0B A13602.0I A13602.30
A13602.0BEA 58.5 EUR
A13602.0B A13602.0I A13602.30
o-Toluidin ≥99%
o-Toluidin

Purity: 99% CAS No.: 95-53-4 Molecular Formula: C7H9N Formula Weight: 107.16 Flash Point: 85°(185°F) Refractive Index: 1.5720 Odour: Unpleasant Melting Point: -24° Boiling Point: 199-200° Beilstein Reference: 741981 Merck Reference: 14,9536 UN No.: 1708 MDL No.: MFCD00007730

Formel: C₇H₉N
Molecular Weight: 107.16 g/mol
Siedepunkt: 199…200 °C
Schmelzpunkt: –24 °C
Dichte: 1.004
Flammpunkt: 85 °C (185 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007730
CAS-Nummer: 95-53-4
EINECS: 202-429-0
UN: 1708
ADR: 6.1 II
Merck Index: 12,09674

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR