2-(4-Chlor-3-nitrobenzoyl)benzoesäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A13605.18
A13605.30
A13605.18EA
35.3
EUR
A13605.18
A13605.30
2-(4-Chlor-3-nitrobenzoyl)benzoesäure ≥98%
2-(4-Chlor-3-nitrobenzoyl)benzoesäure
Purity: 98%
CAS No.: 85-54-1
Molecular Formula: C14H8ClNO5
Formula Weight: 305.68
Melting Point: 198-201°
MDL No.: MFCD00007082
Formel:
C₁₄H₈ClNO₅ Molecular Weight: 305.67 g/mol Schmelzpunkt: 198…201 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007082 CAS-Nummer: 85-54-1 EINECS: 201-613-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...