3-(Dimethylamino)benzoesäure 98%

Lieferant: Thermo Fisher Scientific

A13612.14
A13612.14EA 60.3 EUR
A13612.14
3-(Dimethylamino)benzoesäure 98%
3-(Dimethylamino)benzoesäure

Purity: 98% CAS No.: 99-64-9 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Odour: Odourless Melting Point: 148-153° Beilstein Reference: 2208586 MDL No.: MFCD00002497

Formel: C₉H₁₁NO₂
Siedepunkt: 298 °C (1013 hPa)
Schmelzpunkt: 148…153 °C
Dichte: 1,26 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002497
CAS-Nummer: 99-64-9
EINECS: 202-775-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR