4'-tert-Butylacetophenon ≥98%

Lieferant: Thermo Scientific

Synonyme: p-tert-Butylacetophenon, 1-(4-tert-Butylphenyl)ethanon, 4-tert-Butyl-1-acetylbenzol

A13615.03
A13615.03EA 30.5 EUR
A13615.03
4'-tert-Butylacetophenon ≥98%
4'-tert-Butylacetophenon

Purity: 98% CAS No.: 943-27-1 Molecular Formula: C12H16O Formula Weight: 176.26 Flash Point: 100°(212°F) Refractive Index: 1.5200 Melting Point: 17-18° (dimorphic) Boiling Point: 107-108°/5mm MDL No.: MFCD00017256

Formel: C₁₂H₁₆O
Molecular Weight: 176,26 g/mol
Siedepunkt: 107…108 °C (5 mmHg)
Schmelzpunkt: 17…18 °C (dimorph)
Dichte: 0,964
Flammpunkt: >100 °C (212 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00017256
CAS-Nummer: 943-27-1
EINECS: 213-399-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR