5-Bromisatin ≥90%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 5-Brom-2,3-indolindion, 5-Brom-1H-indol-2,3-dion

A13641.18 A13641.22 A13641.36
A13641.18EA 80.2 EUR
A13641.18 A13641.22 A13641.36
5-Bromisatin ≥90%
5-Bromisatin

Purity: 90+% CAS No.: 87-48-9 Molecular Formula: C8H4BrNO2 Formula Weight: 226.03 Melting Point: ca 250° dec. Beilstein Reference: 383760

Formel: C₈H₄BrNO₂
Molecular Weight: 226.03 g/mol
Schmelzpunkt: ∼ 250 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149345
CAS-Nummer: 87-48-9
EINECS: 201-747-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR