1,5-Hexadien ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Hexa-1,5-diene
A13650.14
A13650.22
A13650.36
A13650.14EA
36.6
EUR
A13650.14
A13650.22
A13650.36
1,5-Hexadien ≥98%
1,5-Hexadien
Purity: 98%
CAS No.: 592-42-7
Molecular Formula: CH2=CHCH2CH2CH=CH2
Formula Weight: 82.15
Flash Point: -27°(-16°F)
Refractive Index: 1.4040
Odour: Turpentine-like
Melting Point: -141°
Boiling Point: 58-60°
Beilstein Reference: 1731106
UN No.: 2458
MDL No.: MFCD00008666
Formel:
C₆H₁₀ Molecular Weight: 82.15 g/mol Siedepunkt: 58…60 °C Schmelzpunkt: –141 °C Dichte: 0.692 Flammpunkt: –27 °C (–16 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008666 CAS-Nummer: 592-42-7 EINECS: 209-754-7 UN: 2458 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...