L(+)-Weinsäure ≥99%
Lieferant: Thermo Fisher Scientific
Purity: 99%
CAS No.: 87-69-4
Molecular Formula: C4H6O6
Formula Weight: 150.09
Odour: Odourless
pH: 2.1
Melting Point: 168-170°
Beilstein Reference: 1725147
Merck Reference: 14,9070
Optical Rotation: +12.4° (c=20 in water)
MDL No.: MFCD00064207
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...