≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-(2-Hydroxyethyl)-1-piperazinepropanesulphonic acid, N-(2-Hydroxyethyl)piperazine-N′-(3-propanesulphonic acid), HEPPS, 3-[4-(2-Hydroxyethyl)-1-piperazinyl]propanesulphonic acid, 3-[4-(2-Hydroxyethyl)-1-piperazinyl]-propanesulphonic acid, 4-(2-Hydroxyethyl)piperazine-1-propanesulphonic acid

A13714.14 A13714.22
A13714.14EA 37.2 EUR
A13714.14 A13714.22
≥99%
EPPS (HEPPS, 4-(2-Hydroxyethyl)-1-piperazinpropansulfonsäure)

Purity: 99% CAS No.: 16052-06-5 Molecular Formula: C9H20N2O4S Formula Weight: 252.33 Melting Point: 242° dec. Beilstein Reference: 3957385

Formel: C₉H₂₀N₂O₄S
Schmelzpunkt: 242 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006160
CAS-Nummer: 16052-06-5
EINECS: 240-198-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR