Kinetin 99%
Lieferant: Thermo Fisher Scientific
Purity: 99%
CAS No.: 525-79-1
Molecular Formula: C10H9N5O
Formula Weight: 215.22
Storage Temperature: 2°C to 8°C
Odour: Odourless
Melting Point: ca 270° dec.
Beilstein Reference: 21703
MDL No.: MFCD00075757
Applications: Plant growth accelerator
Formel:
C₁₀H₉N₅O Lagertemperatur: Gefrierschrank |
MDL:
MFCD00075757 CAS-Nummer: 525-79-1 EINECS: 208-382-2 Merck Index: 14,5311 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...