EEDQ (Ethyl-N-(2-ethoxy-1,2-dihydrochinolin)carboxylat) ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 99%
CAS No.: 16357-59-8
Molecular Formula: C14H17NO3
Formula Weight: 247.29
Storage Temperature: 2°C to 8°C
Melting Point: 63-66°
Boiling Point: 125-128°/0.1mm
Beilstein Reference: 533048
Merck Reference: 14,3518
MDL No.: MFCD00006703
Formel:
C₁₄H₁₇NO₃ Siedepunkt: 125…128 °C (0,1 mmHg) Schmelzpunkt: 63…66 °C |
MDL:
MFCD00006703 CAS-Nummer: 16357-59-8 EINECS: 240-418-2 Merck Index: 14,3518 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...