Geraniol ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 97%
CAS No.: 106-24-1
Molecular Formula: C10H18O
Formula Weight: 154.25
Flash Point: 101°(213°F)
Refractive Index: 1.4760
Odour: Floral
Melting Point: -15°
Boiling Point: 114-115°/12mm
Beilstein Reference: 1722456
Merck Reference: 14,4403
MDL No.: MFCD00002917
Formel:
C₁₀H₁₈O Siedepunkt: 114…115 °C (12 mmHg) Schmelzpunkt: –15 °C Dichte: 0,884 Flammpunkt: 101 °C (213 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002917 CAS-Nummer: 106-24-1 EINECS: 203-377-1 Merck Index: 14,4403 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...