2-Fluorbenzaldehyd ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 97%
CAS No.: 446-52-6
Molecular Formula: C7H5FO
Formula Weight: 124.11
Flash Point: 55°(131°F)
Refractive Index: 1.5220
Odour: Odourless
Melting Point: -45°
Boiling Point: 172-174°
Beilstein Reference: 507155
UN No.: 1989
MDL No.: MFCD00003302
Formel:
C₇H₅FO Siedepunkt: 172…174 °C Schmelzpunkt: –45 °C Dichte: 1,180 Flammpunkt: 55 °C (131 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003302 CAS-Nummer: 446-52-6 EINECS: 207-171-2 UN: 1989 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...