Phenyltrimethoxysilan ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

A13827.14 A13827.22 A13827.36
A13827.14EA 27.3 EUR
A13827.14 A13827.22 A13827.36
Phenyltrimethoxysilan ≥97%
Phenyltrimethoxysilan

Purity: 97% CAS No.: 2996-92-1 Molecular Formula: C6H5Si(OCH3)3 Formula Weight: 198.30 Flash Point: 86°(186°F) Refractive Index: 1.4735 Melting Point: -25° Boiling Point: 210-212° Beilstein Reference: 2937896 MDL No.: MFCD00025689

Formel: C₉H₁₄O₃Si
Siedepunkt: 210…212 °C
Schmelzpunkt: –25 °C
Dichte: 1,070
Flammpunkt: 86 °C (186 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00025689
CAS-Nummer: 2996-92-1
EINECS: 221-066-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR