1,2-Phenylendiessigsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A13851.06 A13851.14
A13851.06EA 19.1 EUR
A13851.06 A13851.14
1,2-Phenylendiessigsäure ≥98%
1,2-Phenylendiessigsäure

Purity: 98+% CAS No.: 7500-53-0 Molecular Formula: C10H10O4 Formula Weight: 194.19 Odour: Odourless Melting Point: 150-152° Beilstein Reference: 2616747 MDL No.: MFCD00004329

Formel: C₁₀H₁₀O₄
Schmelzpunkt: 150…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004329
CAS-Nummer: 7500-53-0
EINECS: 231-355-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR