1-Fluor-2-nitrobenzol ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2-Fluornitrobenzol

A13852.0B A13852.18 A13852.30
A13852.0BEA 314 EUR
A13852.0B A13852.18 A13852.30
1-Fluor-2-nitrobenzol ≥99%
1-Fluor-2-nitrobenzol

Purity: 99% CAS No.: 1493-27-2 Molecular Formula: C6H4FNO2 Formula Weight: 141.10 Flash Point: 94°(201°F) Physical Form: Liquid Density: 1.340 Refractive Index: 1.5317 Melting Point: -8° Boiling Point: 215° Beilstein Reference: 607262 UN No.: 2810 MDL No.: MFCD00007048

Formel: C₆H₄FNO₂
Molecular Weight: 141.1 g/mol
Siedepunkt: 215 °C
Schmelzpunkt: –8 °C
Dichte: 1.340
Flammpunkt: 94 °C (201 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007048
CAS-Nummer: 1493-27-2
EINECS: 216-088-0
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR