1-Fluor-2-nitrobenzol ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2-Fluornitrobenzol
A13852.0B
A13852.18
A13852.30
A13852.0BEA
314
EUR
A13852.0B
A13852.18
A13852.30
1-Fluor-2-nitrobenzol ≥99%
1-Fluor-2-nitrobenzol
Purity: 99%
CAS No.: 1493-27-2
Molecular Formula: C6H4FNO2
Formula Weight: 141.10
Flash Point: 94°(201°F)
Physical Form: Liquid
Density: 1.340
Refractive Index: 1.5317
Melting Point: -8°
Boiling Point: 215°
Beilstein Reference: 607262
UN No.: 2810
MDL No.: MFCD00007048
Formel:
C₆H₄FNO₂ Molecular Weight: 141.1 g/mol Siedepunkt: 215 °C Schmelzpunkt: –8 °C Dichte: 1.340 Flammpunkt: 94 °C (201 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007048 CAS-Nummer: 1493-27-2 EINECS: 216-088-0 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...