(4-Hydroxy-3-nitrophenyl)essigsäure ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-(4-Hydroxy-3-nitrophenyl)acetic acid
A13858.06
A13858.14
A13858.06EA
128
EUR
A13858.06
A13858.14
(4-Hydroxy-3-nitrophenyl)essigsäure ≥99%
(4-Hydroxy-3-nitrophenyl)essigsäure
Purity: 99%
CAS No.: 10463-20-4
Molecular Formula: C8H7NO5
Formula Weight: 197.15
Melting Point: 146-148°
Beilstein Reference: 2696044
Formel:
(C₆H₃(OH)(NO₂))CH₂COOH Schmelzpunkt: 146…148 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007122 CAS-Nummer: 10463-20-4 EINECS: 233-948-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...