N-Hydroxyphthalimid ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: N-Hydroxyphthalsäureimid, N-Hydroxy-1,2-benzoldicarbonsäureimid

A13862.22 A13862.36
A13862.22EA 35.4 EUR
A13862.22 A13862.36
N-Hydroxyphthalimid ≥98%
N-Hydroxyphthalimid

Purity: 98+% CAS No.: 524-38-9 Molecular Formula: C8H5NO3 Formula Weight: 163.13 Melting Point: ca 233° dec. Beilstein Reference: 131208 Merck Reference: 14,4838 MDL No.: MFCD00005891

Formel: C₈H₅NO₃
Molecular Weight: 163.13 g/mol
Siedepunkt: 412
Schmelzpunkt: ∼ 233 °C (Zers.)
Dichte: 1.58 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005891
CAS-Nummer: 524-38-9
EINECS: 208-358-1
Merck Index: 13,04864

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR