3,3',5,5'-Tetramethylbenzidin (TMB) ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: TMB

A13868.03 A13868.06 A13868.14
A13868.03EA 48.4 EUR
A13868.03 A13868.06 A13868.14
3,3',5,5'-Tetramethylbenzidin (TMB) ≥98%
3,3',5,5'-Tetramethylbenzidin (TMB)

Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes; particularly useful in ELISA techniques, blue result can be read spectrophotometrically at 370 or 650 nm3,3',5,5'-Tetramethylbenzidine is used as a clinical reagent for testing blood.


It is also a chromogenic substrate which is used in staining procedures in immunohistochemistry. Further, it is used as a visualizing reagent in enzyme-linked immunosorbent assays. In addition to this, it is used as a replacement for carcinogenic compounds such as benzidine and o-phenylenediamine.

Formel: C₁₆H₂₀N₂
Siedepunkt: 402 °C (1013 hPa)
Schmelzpunkt: 168…171 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007748
CAS-Nummer: 54827-17-7
EINECS: 259-364-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR