trans-4-Methoxycinnamic acid 98%

Lieferant: Thermo Fisher Scientific
Achtung

A13870.18 A13870.09 A13870.30
A13870.18EA 73.9 EUR
A13870.18 A13870.09 A13870.30
trans-4-Methoxycinnamic acid 98%
trans-4-Methoxycinnamic acid

Purity: 98% CAS No.: 943-89-5 Molecular Formula: C10H10O3 Formula Weight: 178.19 Melting Point: 171-175° Beilstein Reference: 510468

Formel: C₁₀H₁₀O₃
Siedepunkt: 317 °C (1013 hPa)
Schmelzpunkt: 171…175 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004398
CAS-Nummer: 830-09-1
EINECS: 213-405-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR