N,N'-Bis(salicyliden)ethylendiamin 98%

Lieferant: Thermo Fisher Scientific
Achtung

A13885.14 A13885.22 A13885.36
A13885.14EA 46 EUR
A13885.14 A13885.22 A13885.36
N,N'-Bis(salicyliden)ethylendiamin 98%
N,N'-Bis(salicyliden)ethylendiamin

Purity: 98% CAS No.: 94-93-9 Molecular Formula: C16H16N2O2 Formula Weight: 268.32 Melting Point: 129-131° Beilstein Reference: 535296 Merck Reference: 14,8322 MDL No.: MFCD00002244

Formel: C₁₆H₁₆N₂O₂
Schmelzpunkt: 129…131 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002244
CAS-Nummer: 94-93-9
EINECS: 202-376-3
Merck Index: 14,8322

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR