N,N'-Bis(salicyliden)ethylendiamin 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A13885.14
A13885.22
A13885.36
A13885.14EA
46
EUR
A13885.14
A13885.22
A13885.36
N,N'-Bis(salicyliden)ethylendiamin 98%
N,N'-Bis(salicyliden)ethylendiamin
Purity: 98%
CAS No.: 94-93-9
Molecular Formula: C16H16N2O2
Formula Weight: 268.32
Melting Point: 129-131°
Beilstein Reference: 535296
Merck Reference: 14,8322
MDL No.: MFCD00002244
Formel:
C₁₆H₁₆N₂O₂ Schmelzpunkt: 129…131 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002244 CAS-Nummer: 94-93-9 EINECS: 202-376-3 Merck Index: 14,8322 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...