trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid (trans-) ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

A13890.14 A13890.22 A13890.36
A13890.14EA 55.6 EUR
A13890.14 A13890.22 A13890.36
trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid (trans-) ≥99%
trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid

Purity: 99% CAS No.: 537-98-4 Molecular Formula: C10H10O4 Formula Weight: 194.19 Melting Point: 170-175° Beilstein Reference: 1371483 Merck Reference: 14,4062 MDL No.: MFCD00004400

Formel: C₁₀H₁₀O₄
Schmelzpunkt: 170…175 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004400
CAS-Nummer: 537-98-4
EINECS: 208-679-7
Merck Index: 14,4062

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR