ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Mononatriumsalz, oxidierte Form (NADP-Na)

Lieferant: PanReac AppliChem

Synonyme: NADP Mononatriumsalz, ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Mononatriumsalz, NADP-Na, Nadidphosphat Mononatriumsalz, Nicotinamid-Adenin-Dinucleotid-Phosphat Mononatriumsalz

A1394.0001 A1394.0005 A1394.9050
A1394.0001EA 169 EUR
A1394.0001 A1394.0005 A1394.9050
ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Mononatriumsalz, oxidierte Form (NADP-Na)
ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Mononatriumsalz, oxidierte Form(NADP-Na)
Formel: C₂₁H₂₇N₇NaO₁₇P₃
Molecular Weight: 765.39 g/mol
MDL: MFCD00036973
CAS-Nummer: 1184-16-3
EINECS: 214-664-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR