Thiophen ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

A13941.0E A13941.18 A13941.30 A13941.36
A13941.0EEA 294 EUR
A13941.0E A13941.18 A13941.30 A13941.36
Thiophen ≥99%
Thiophen

Purity: 99% CAS No.: 110-02-1 Molecular Formula: C4H4S Formula Weight: 84.14 Flash Point: -9°(16°F) Refractive Index: 1.5280 Odour: Stench Melting Point: -38° Boiling Point: 83-85° Beilstein Reference: 103222 Merck Reference: 14,9353 UN No.: 2414 MDL No.: MFCD00005413

Formel: C₄H₄S
Siedepunkt: 83…85 °C
Schmelzpunkt: –38 °C
Dichte: 1,066
Flammpunkt: –9 °C (16 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005413
CAS-Nummer: 110-02-1
EINECS: 203-729-4
UN: 2414
ADR: 3,II
Merck Index: 14,9353

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR