1,2-Diamino-4-chloro-5-fluorobenzene 98%

Lieferant: Thermo Fisher Scientific
Gefahr

A13953.03 A13953.06
A13953.03EA 47.7 EUR
A13953.03 A13953.06
1,2-Diamino-4-chloro-5-fluorobenzene 98%
1,2-Diamino-4-chloro-5-fluorobenzene

Purity: 98% CAS No.: 139512-70-2 Molecular Formula: C6H6ClFN2 Formula Weight: 160.58 Melting Point: 110-114° UN No.: UN2811 MDL No.: MFCD00042485

Formel: C₆H₆ClFN₂
Schmelzpunkt: 110…114 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00042485
CAS-Nummer: 139512-70-2
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR