5-Hydroxy-L-tryptophan Hydrat 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A13954.03
A13954.03EA
34.1
EUR
A13954.03
5-Hydroxy-L-tryptophan Hydrat 98%
5-Hydroxy-L-tryptophan Hydrat
For routes to 2-bromo-5-hydroxytryptophan derivatives, see: Synthesis, 28 (1996).
Formel:
C₁₁H₁₂N₂O₃·xH₂O Schmelzpunkt: 258 °C (Zers.) Lagertemperatur: Kühlschrank |
MDL:
MFCD00064341 CAS-Nummer: 4350-09-8 EINECS: 224-411-1 UN: 2811 ADR: 6.1,III Merck Index: 14,4847 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...