Ammoniumcarbonat (∼30% NH₃ )

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Ammoniumcarbonat, Hirschhornsalz

A13955.0B A13955.0I
A13955.0BEA 35.9 EUR
A13955.0B A13955.0I
Ammoniumcarbonat (∼30% NH₃ )
Ammoniumcarbonat (enthält Ammoniumcarbamat)

CAS No.: 506-87-6 Molecular Formula: CH8N2O3 Formula Weight: 96.09 Odour: Ammonia-like Melting Point: 60° subl. Beilstein Reference: 3627235 Merck Reference: 14,508 MDL No.: MFCD00010890

Formel: (NH₄)₂CO₃
Molecular Weight: 96.09 g/mol
Schmelzpunkt: 60 °C (Subl.)
Dichte: 1.5 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010890
CAS-Nummer: 506-87-6
EINECS: 208-058-0
Merck Index: 13,00508

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR