2-Amino-4-nitrophenol 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Hydroxy-5-nitroanilin

A13964.14
A13964.14EA 77.4 EUR
A13964.14
2-Amino-4-nitrophenol 98%
2-Amino-4-nitrophenol

Purity: 98% CAS No.: 99-57-0 Molecular Formula: C6H6N2O3 Formula Weight: 154.13 Melting Point: 141-145° Beilstein Reference: 776533

Formel: C₆H₆N₂O₃
Schmelzpunkt: 141…145 °C
Dichte: 1,66 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007695
CAS-Nummer: 99-57-0
EINECS: 202-767-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR