1,5-Pentandiol ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Pentamethylenglykol, Pentamethylenglycol, 1,5-Dihydroxypentan

A13998.0E A13998.22 A13998.36
A13998.0EEA 183 EUR
A13998.0E A13998.22 A13998.36
1,5-Pentandiol ≥97%
1,5-Pentandiol

Purity: 97% CAS No.: 111-29-5 Molecular Formula: C5H12O2 Formula Weight: 104.15 Flash Point: 129°(264°F) Refractive Index: 1.4500 Melting Point: -16° Boiling Point: 241-242° Beilstein Reference: 1560130 Merck Reference: 14,7117 MDL No.: MFCD00002978

Formel: C₅H₁₂O₂
Molecular Weight: 104.15 g/mol
Siedepunkt: 241…242 °C
Schmelzpunkt: –16 °C
Dichte: 0.992
Flammpunkt: 129 °C (264 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002978
CAS-Nummer: 111-29-5
EINECS: 203-854-4
Merck Index: 14,7117

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR