2,5-Difluorbenzonitril ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

A14014.06
A14014.06EA 48.1 EUR
A14014.06
2,5-Difluorbenzonitril ≥98%
2,5-Difluorbenzonitril

Purity: 98+% CAS No.: 64248-64-2 Molecular Formula: C7H3F2N Formula Weight: 139.11 Flash Point: 77°(170°F) Melting Point: 31-35° Boiling Point: 188° Beilstein Reference: 2085640 UN No.: 3439 MDL No.: MFCD00001777

Formel: C₇H₃F₂N
Molecular Weight: 139.1 g/mol
Siedepunkt: 188 °C
Schmelzpunkt: 31…35 °C
Flammpunkt: 77 °C (170 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00001777
CAS-Nummer: 64248-64-2
EINECS: 264-753-9
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR