Chloressigsäureanhydrid ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 97%
CAS No.: 541-88-8
Molecular Formula: (ClCH2CO)2O
Formula Weight: 170.98
Flash Point: 143°(289°F)
Melting Point: 56-59°
Boiling Point: 110-112°/10mm
Solubility: Freely soluble in ether, chloroform. Slightly soluble in benzene. Insoluble in cold ligroin
Beilstein Reference: 774533
Merck Reference: 14,2113
UN No.: 3261
MDL No.: MFCD00000929
Formel:
C₄H₄Cl₂O₃ Siedepunkt: 110…112 °C (10 mmHg) Schmelzpunkt: 56…59 °C Dichte: 1,5497 g/cm³ (20 °C) Flammpunkt: 143 °C (289 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000929 CAS-Nummer: 541-88-8 EINECS: 208-794-2 UN: 3261 ADR: 8,II Merck Index: 14,2113 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...