2-Methoxybenzylalkohol ≥99%
Lieferant: Thermo Fisher Scientific
Synonyme:
(2-Methoxyphenyl)methanol, 2-(Hydroxymethyl)anisol
A14016.14
A14016.22
A14016.14EA
50.7
EUR
A14016.14
A14016.22
2-Methoxybenzylalkohol ≥99%
2-Methoxybenzylalkohol
Purity: 99%
CAS No.: 612-16-8
Molecular Formula: C8H10O2
Formula Weight: 138.17
Flash Point:
Formel:
C₈H₁₀O₂ Molecular Weight: 138.17 g/mol Siedepunkt: 248…250 °C Dichte: 1.12 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004611 CAS-Nummer: 612-16-8 EINECS: 210-296-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...