Methyl-(triphenylphosphoranyliden)acetat 98%

Lieferant: Thermo Fisher Scientific

Synonyme: Methyl (triphenylphosphoranylidene)acetate, Methyl 2-(triphenylphosphoranylidene)acetate, Methoxycarbonylmethylene-Triphenylphosphorane

A14020.14 A14020.22 A14020.36
A14020.14EA 101 EUR
A14020.14 A14020.22 A14020.36
Methyl-(triphenylphosphoranyliden)acetat 98%
Methyl-2-(triphenylphosphoranylidene)acetate

Purity: 98% CAS No.: 2605-67-6 Molecular Formula: CH3O2CCH=P(C6H5)3 Formula Weight: 334.36 Melting Point: 165-169° Beilstein Reference: 618430 MDL No.: MFCD00008455

Formel: C₂₁H₁₉O₂P
Molecular Weight: 334.36 g/mol
Schmelzpunkt: 165…169 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008455
CAS-Nummer: 2605-67-6
EINECS: 220-018-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR