1,5-Diphenylcarbazid ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

A14060.14 A14060.22 A14060.36
A14060.14EA 65.2 EUR
A14060.14 A14060.22 A14060.36
1,5-Diphenylcarbazid ≥97%
1,5-Diphenylcarbazid

Purity: 97+% CAS No.: 140-22-7 Molecular Formula: C13H14N4O Formula Weight: 242.28 Melting Point: 170-176° Beilstein Reference: 752039 Merck Reference: 14,3321 MDL No.: MFCD00003013

Formel: C₁₃H₁₄N₄O
Siedepunkt: 454 °C (1013 hPa)
Schmelzpunkt: 170…176 °C
Dichte: 1,31 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003013
CAS-Nummer: 140-22-7
EINECS: 205-403-7
Merck Index: 14,3321
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03cf8836a36f1077443ec2e3cfa151621925bd46&vid=a1b2bd876c3fbd9d6ded95c8430e4ec16bf5172b&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR