4-Hydroxybenzylaceton ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

A14064.14 A14064.22 A14064.30
A14064.14EA 51.6 EUR
A14064.14 A14064.22 A14064.30
4-Hydroxybenzylaceton ≥98%
4-Hydroxybenzylaceton

Purity: 98% CAS No.: 5471-51-2 Molecular Formula: C10H12O2 Formula Weight: 164.20 Melting Point: 80-84° Beilstein Reference: 776080

Formel: C₁₀H₁₂O₂
Siedepunkt: 292 °C (1013 hPa)
Schmelzpunkt: 80…84 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002394
CAS-Nummer: 5471-51-2
EINECS: 226-806-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR