Glycinmethylester Hydrochlorid (H-Gly-OMe.HCl) ≥99%

Lieferant: Thermo Fisher Scientific

Synonyme: Methyl glycinate hydrochloride, H-Gly-OMe.HCl, Glycine Methyl Ester HCl

A14083.0E A14083.22 A14083.36
A14083.0EEA 444 EUR
A14083.0E A14083.22 A14083.36
Glycinmethylester Hydrochlorid (H-Gly-OMe.HCl) ≥99%
Glycinmethylester Hydrochlorid (H-Gly-OMe.HCl)

Purity: 99% CAS No.: 5680-79-5 Molecular Formula: H2NCH2CO2CH3.HCl Formula Weight: 125.56 Melting Point: ca 175° dec. Beilstein Reference: 3593644

Formel: C₃H₈ClNO₂
Schmelzpunkt: ∼ 175 °C (Zersetzung)
Dichte: 1,000
Lagertemperatur: Raumtemperatur
MDL: MFCD00012870
CAS-Nummer: 5680-79-5
EINECS: 227-139-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR