Geranylbromid 96%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 96%
CAS No.: 6138-90-5
Molecular Formula: C10H17Br
Formula Weight: 217.16
Storage Temperature: 2°C to 8°C
Flash Point: 95°(203°F)
Refractive Index: 1.5040
Boiling Point: 99-101°/10mm
Beilstein Reference: 1422412
MDL No.: MFCD00000243
Formel:
C₁₀H₁₇Br Siedepunkt: 99…101 °C (10 mmHg) Dichte: 1,094 Flammpunkt: 95 °C (203 °F) |
MDL:
MFCD00000243 CAS-Nummer: 6138-90-5 EINECS: 228-123-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...