1H-Pyrazol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A14186.14 A14186.22
A14186.14EA 50 EUR
A14186.14 A14186.22
1H-Pyrazol ≥98%
1H-Pyrazol

Purity: 98% CAS No.: 288-13-1 Molecular Formula: C3H4N2 Formula Weight: 68.08 Odour: Unpleasant Melting Point: 66-70° Boiling Point: 186-188° Beilstein Reference: 103775 Merck Reference: 14,7960 MDL No.: MFCD00005234

Formel: C₃H₄N₂
Siedepunkt: 186…188 °C
Schmelzpunkt: 66…70 °C
Dichte: 1,34 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005234
CAS-Nummer: 288-13-1
EINECS: 206-017-1
Merck Index: 14,7960

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR