1,3-Dihydroxyaceton dimer ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A14189.0B
A14189.18
A14189.30
A14189.0BEA
294
EUR
A14189.0B
A14189.18
A14189.30
1,3-Dihydroxyaceton dimer ≥97%
1,3-Dihydroxyaceton dimer
Purity: 97%
CAS No.: 62147-49-3
Molecular Formula: C6H12O6
Formula Weight: 180.16
Storage Temperature: 2°C to 8°C
Melting Point: ca 75-82°
Beilstein Reference: 112910
Merck Reference: 14,3182
MDL No.: MFCD00051019
Formel:
C₆H₁₂O₆ Schmelzpunkt: ∼ 75…82 °C |
MDL:
MFCD00051019 CAS-Nummer: 62147-49-3 EINECS: 612-947-2 Merck Index: 14,3182 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...